1,1-Bis((2S,5S)-2,5-dimethylphospholano)ferrocene(cyclooctadiene)rhodium(I) 98%

Supplier: Ambeed
Warning

A571891-100MG A571891-1G A571891-250MG
77254-064EA 144.2 USD
77254-064 77254-066 77254-068
1,1-Bis((2S,5S)-2,5-dimethylphospholano)ferrocene(cyclooctadiene)rhodium(I) 98%
1,1-Bis((2S,5S)-2,5-dimethylphospholano)ferrocene(cyclooctadiene)rhodium(I)

Precaución: for Research Use Only

Formula: C₃₀H₄₄FeP₂Rh
MW: 626.17 g/mol
MDL Number: MFCD01631281
CAS Number: 854920-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR