Pyridoxal hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride, Pyridoxal HCl

A571882-25G A571882-5G A571882-100G
77254-060EA 86 USD
77254-060 77254-062 77747-184
Pyridoxal hydrochloride 98%
Pyridoxal hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₃
MW: 203.62 g/mol
MDL Number: MFCD00012809
CAS Number: 65-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR