Methyl-4-iodo-2-methoxybenzoate 95%

Supplier: Ambeed
Warning

Synonyms: 5-Iodo-2-(methoxycarbonyl)anisole, Methyl 4-iodo-2-methoxybenzoate, 4-Iodo-2-methoxybenzoic acid methyl ester, Methyl-4-iodo-2-methoxybenzoate, Methyl 4-iodo-o-anisate, Methyl 3-iodo-4-methoxybenzoate

A571861-1G A571861-25G A571861-5G
77280-104EA 14 USD
77280-104 77280-106 77280-108
Methyl-4-iodo-2-methoxybenzoate 95%
Methyl-4-iodo-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉IO₃
MW: 292.07
MDL Number: MFCD00052925
CAS Number: 35387-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR