Dodecylamine hydrobromide 97%

Supplier: Ambeed
Warning

A571710-5G A571710-1G
77254-058EA 47 USD
77254-058 77747-182
Dodecylamine hydrobromide 97%
Dodecylamine hydrobromide

Precaución: for Research Use Only

Formula: C₁₂H₂₇N·HBr
MDL Number: MFCD30543762
CAS Number: 26204-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR