2-Hydroxy-5-nitropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Nitro-2-pyridinol

A571098-100G A571098-10G A571098-25G A571098-500G A571098-5G
77292-606EA 41.2 USD
77292-606 77292-608 77292-610 77292-612 77292-614
2-Hydroxy-5-nitropyridine 98%
2-Hydroxy-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
MDL Number: MFCD00006276
CAS Number: 5418-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR