Fevipiprant 99+%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Methyl-1-(4-(methylsulphonyl)-2-(trifluoromethyl)benzyl)-1H-pyrrolo[2,3-b]pyridin-3-yl)acetic acid, NVP-QAW039, QAW039

A570568-1MG A570568-25MG A570568-5MG
77266-440EA 12 USD
77266-440 77266-442 77266-444
Fevipiprant 99+%
Fevipiprant

Precaución: for Research Use Only

Formula: C₁₉H₁₇F₃N₂O₄S
MDL Number: MFCD28963975
CAS Number: 872365-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR