2,6-Difluoro-4-iodoaniline 97%

Supplier: Ambeed
Warning

A570212-100G A570212-10G A570212-1G A570212-250MG A570212-25G A570212-5G
77254-038EA 203.94 USD
77254-038 77254-040 77254-042 77254-044 77254-046 77254-048
2,6-Difluoro-4-iodoaniline 97%
2,6-Difluoro-4-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄F₂IN
MW: 255.0039 g/mol
MDL Number: MFCD07774189
CAS Number: 141743-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR