(S)-1-(1-(Imidazo[1,2-a]pyridin-6-yl)ethyl)-6-(1-methyl-1H-pyrazol-4-yl)-1H-[1,2,3]triazolo[4,5-b]pyrazine 98%

Supplier: Ambeed
Warning

Synonyms: Savolitinib

A569590-10MG A569590-1MG A569590-50MG A569590-5MG A569590-100MG
77242-234EA 101.97 USD
77242-234 77242-236 77242-238 77242-240 77747-174
(S)-1-(1-(Imidazo[1,2-a]pyridin-6-yl)ethyl)-6-(1-methyl-1H-pyrazol-4-yl)-1H-[1,2,3]triazolo[4,5-b]pyrazine 98%
(S)-1-(1-(Imidazo[1,2-a]pyridin-6-yl)ethyl)-6-(1-methyl-1H-pyrazol-4-yl)-1H-[1,2,3]triazolo[4,5-b]pyrazine

Precaución: for Research Use Only

Formula: C₁₇H₁₅N₉
MW: 345.36 g/mol
MDL Number: MFCD28359096
CAS Number: 1313725-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR