2-(3-Aminophenyl)acetic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Aminophenylacetic acid
A569400-1G
A569400-5G
77280-092EA
12.36
USD
77280-092
77749-172
2-(3-Aminophenyl)acetic acid 98%
2-(3-Aminophenyl)acetic acid
Precaución: for Research Use Only
Formula:
C₈H₉NO₂ MW: 151.1626 g/mol |
MDL Number:
MFCD00075058 CAS Number: 14338-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...