2-Amino-6-methoxybenzothiazole 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxybenzothiazol-2-ylamine

A569353-1000G A569353-100G A569353-500G A569353-25G
77254-032EA 78.28 USD
77254-032 77254-034 77254-036 77307-844
2-Amino-6-methoxybenzothiazole 97%
2-Amino-6-methoxybenzothiazole

Precaución: for Research Use Only

Formula: C₈H₈N₂OS
MW: 180.23 g/mol
MDL Number: MFCD00005787
CAS Number: 1747-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR