2-(2-Pyridyl)benzimidazole 98%

Supplier: Ambeed
Warning

A568288-10G A568288-1G A568288-25G A568288-5G A568288-100G
77242-226EA 27 USD
77242-226 77242-228 77242-230 77242-232 77747-170
2-(2-Pyridyl)benzimidazole 98%
2-(2-Pyridyl)benzimidazole

Precaución: for Research Use Only

Formula: C₁₂H₉N₃
MDL Number: MFCD00005586
CAS Number: 1137-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR