6'-(Diethylamino)-2'-((2,4-dimethylphenyl)amino)-3'-methyl-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one 98%

Supplier: Ambeed
Warning

A568287-100G A568287-25G A568287-500G
77242-222EA 30 USD
77242-222 77242-224 77617-402
6'-(Diethylamino)-2'-((2,4-dimethylphenyl)amino)-3'-methyl-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one 98%
6'-(Diethylamino)-2'-((2,4-dimethylphenyl)amino)-3'-methyl-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

Precaución: for Research Use Only

Formula: C₃₃H₃₂N₂O₃
MDL Number: MFCD08703297
CAS Number: 36431-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR