3,4-Dihydroxypyridine 95%

Supplier: Ambeed
Warning

A568217-100MG A568217-250MG A568217-5G A568217-1G
77280-082EA 22 USD
77280-082 77280-084 77280-086 77617-400
3,4-Dihydroxypyridine 95%
3,4-Dihydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₅NO₂
MDL Number: MFCD01691806
CAS Number: 10182-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR