4-(4-Chlorophenyl)morpholine 98%

Supplier: Ambeed
Danger

A568165-5G A568165-1G
77292-592EA 49 USD
77292-592 77749-170
4-(4-Chlorophenyl)morpholine 98%
4-(4-Chlorophenyl)morpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO
MDL Number: MFCD09038510
CAS Number: 70291-67-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR