2-Bromo-5-(4-morpholino)pyridine 96%
Supplier: Ambeed
|
Warning
|
A567997-1G
A567997-25G
A567997-5G
77292-590EA
9
USD
77292-590
77551-714
77569-092
2-Bromo-5-(4-morpholino)pyridine 96%
2-Bromo-5-(4-morpholino)pyridine
Precaución: For research use only.
Formula:
C₉H₁₁BrN₂O |
MDL Number:
MFCD02681930 CAS Number: 200064-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...