4-(Difluoromethoxy)benzenesulfonamide 98%

Supplier: Ambeed
Warning

A567659-10G A567659-1G A567659-25G
77292-582EA 134 USD
77292-582 77292-584 77292-586
4-(Difluoromethoxy)benzenesulfonamide 98%
4-(Difluoromethoxy)benzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₇F₂NO₃S
MDL Number: MFCD03407973
CAS Number: 874781-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR