4-(2-(Dimethylamino)ethylcarbamoyl)phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Danger

Synonyms: N-(2-Dimethylaminoethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide, 4-(N-[2-(Dimethylamino)ethyl]carbamoyl)benzeneboronic acid pinacol ester

A566660-250MG A566660-5G A566660-1G
77292-578EA 14 USD
77292-578 77292-580 77749-167
4-(2-(Dimethylamino)ethylcarbamoyl)phenylboronic acid pinacol ester 95%
4-(2-(Dimethylamino)ethylcarbamoyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₇BN₂O₃
MDL Number: MFCD05864307
CAS Number: 832114-11-1
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR