1-Allyl-3-methylimidazolium bis(trifluoromethanesulfonyl)imide 98%

Supplier: Ambeed
Warning

Synonyms: 1-Allyl-3-methylimidazolium bis((trifluoromethyl)sulphonyl)imide

A566489-1G A566489-25G A566489-5G
77266-406EA 7 USD
77266-406 77266-408 77266-410
1-Allyl-3-methylimidazolium bis(trifluoromethanesulfonyl)imide 98%
1-Allyl-3-methylimidazolium bis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₉H₁₁F₆N₃O₄S₂
MW: 403.31 g/mol
MDL Number: MFCD11109343
CAS Number: 655249-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR