(3-Tolyloxy)acetic acid 98%

Supplier: Ambeed
Warning

A566478-1G A566478-25G A566478-5G
77280-068EA 10 USD
77280-068 77280-070 77280-072
(3-Tolyloxy)acetic acid 98%
(3-Tolyloxy)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄(CH₃))OCH₂COOH
MDL Number: MFCD00014358
CAS Number: 1643-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR