1-(2-Aminoethyl)pyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A566459-100MG A566459-250MG
77254-014EA 75.19 USD
77254-014 77254-016
1-(2-Aminoethyl)pyrimidine-2,4(1H,3H)-dione 97%
1-(2-Aminoethyl)pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
MDL Number: MFCD16683348
CAS Number: 34386-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR