[1,1'-Biphenyl]-4,4'-diyl diacrylate 97%

Supplier: Ambeed
Warning

A566309-1G A566309-250MG A566309-5G
77242-210EA 54.59 USD
77242-210 77242-212 77594-728
[1,1'-Biphenyl]-4,4'-diyl diacrylate 97%
[1,1'-Biphenyl]-4,4'-diyl diacrylate

Precaución: for Research Use Only

Formula: C₁₈H₁₄O₄
MW: 294.30 g/mol
MDL Number: MFCD30187307
CAS Number: 84948-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR