2,4-Dichloro-5,6-dihydrobenzo[h]quinazoline 97%

Supplier: Ambeed
Warning

A566191-100MG A566191-1G A566191-250MG
77242-194EA 159.65 USD
77242-194 77242-196 77242-198
2,4-Dichloro-5,6-dihydrobenzo[h]quinazoline 97%
2,4-Dichloro-5,6-dihydrobenzo[h]quinazoline

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂N₂
MW: 251.11 g/mol
CAS Number: 1186410-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR