1,1-Diphenyl-2-thiourea 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
1,1-Diphenylthiourea
A566150-1G
A566150-250MG
A566150-5G
77242-188EA
67.98
USD
77242-188
77242-190
77242-192
1,1-Diphenyl-2-thiourea 97%
1,1-Diphenyl-2-thiourea
Precaución: for Research Use Only
Formula:
C₁₃H₁₂N₂S MW: 228.3100 g/mol |
MDL Number:
MFCD00059151 CAS Number: 3898-08-6 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...