2-Bromobenzo[d]thiazole-6-carboxamide 97%

Supplier: Ambeed
Danger

A566019-100MG A566019-1G A566019-250MG
77242-182EA 93.73 USD
77242-182 77242-184 77242-186
2-Bromobenzo[d]thiazole-6-carboxamide 97%
2-Bromobenzo[d]thiazole-6-carboxamide

Precaución: for Research Use Only

Formula: C₈H₅BrN₂OS
MW: 257.10 g/mol
MDL Number: MFCD26678975
CAS Number: 1502927-80-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR