2-Bromo-1H-imidazole-4,5-dicarbonitrile 97%

Supplier: Ambeed
Warning

A565667-100MG A565667-1G A565667-250MG A565667-5G
77266-398EA 40 USD
77266-398 77266-400 77266-402 77266-404
2-Bromo-1H-imidazole-4,5-dicarbonitrile 97%
2-Bromo-1H-imidazole-4,5-dicarbonitrile

Precaución: for Research Use Only

Formula: C₅HBrN₄
MDL Number: MFCD00174288
CAS Number: 50847-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR