Methyl 1-methylpyrrole-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methyl-1H-pyrrole-3-carboxylate

A565338-1G A565338-5G
77242-178EA 116.39 USD
77242-178 77242-180
Methyl 1-methylpyrrole-3-carboxylate 97%
Methyl 1-methylpyrrole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.152 g/mol
MDL Number: MFCD19981621
CAS Number: 40611-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR