2-Methyl-1,3-propanediol 95%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dihydroxy-2-methylpropane

A564920-100G A564920-25G
77266-396EA 8.24 USD
77266-396 77749-166
2-Methyl-1,3-propanediol 95%
2-Methyl-1,3-propanediol

Precaución: for Research Use Only

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
MDL Number: MFCD00082586
CAS Number: 2163-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR