(2-Amino-4,5-dimethoxyphenyl)(phenyl)methanone 97%

Supplier: Ambeed
Warning

A564918-1G A564918-250MG
77242-174EA 219.39 USD
77242-174 77242-176
(2-Amino-4,5-dimethoxyphenyl)(phenyl)methanone 97%
(2-Amino-4,5-dimethoxyphenyl)(phenyl)methanone

Precaución: for Research Use Only

Formula: C₁₅H₁₅NO₃
MW: 257.28 g/mol
MDL Number: MFCD06739588
CAS Number: 39996-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR