1-(4-Bromophenyl)-1,2,2-triphenylethylene 98%

Supplier: Ambeed
Warning

A564882-100G A564882-100MG A564882-10G A564882-1G A564882-250MG A564882-5G A564882-25G
77242-162EA 622.99 USD
77242-162 77242-164 77242-166 77242-168 77242-170 77242-172 77747-165
1-(4-Bromophenyl)-1,2,2-triphenylethylene 98%
1-(4-Bromophenyl)-1,2,2-triphenylethylene

Precaución: for Research Use Only

Formula: C₂₆H₁₉Br
MDL Number: MFCD20257904
CAS Number: 34699-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR