2,4-Dimethylthiazole 96%

Supplier: Ambeed
Danger

A564732-10G A564732-5G A564732-25G
77292-564EA 7.21 USD
77292-564 77292-566 77572-796
2,4-Dimethylthiazole 96%
2,4-Dimethylthiazole

Precaución: for Research Use Only

Formula: C₅H₇NS
MW: 113.18 g/mol
MDL Number: MFCD00014509
CAS Number: 541-58-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR