Phenyl(1H-pyrrol-3-yl)methanone 95%

Supplier: Ambeed
Warning

Synonyms: [(1H-Pyrrol-3-yl)carbonyl]benzene, 3-Benzoyl-1H-pyrrole

A564596-1G A564596-250MG
77280-066EA 85.49 USD
77280-066 77749-164
Phenyl(1H-pyrrol-3-yl)methanone 95%
Phenyl(1H-pyrrol-3-yl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₉NO
MW: 171.195 g/mol
MDL Number: MFCD02179582
CAS Number: 7126-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR