2-(4-Bromophenyl)benzoxazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Bromophenyl)benzo[d]oxazole

A564388-250MG A564388-5G A564388-1G
77242-146EA 5.15 USD
77242-146 77242-148 77747-163
2-(4-Bromophenyl)benzoxazole 98%
2-(4-Bromophenyl)benzoxazole

Precaución: for Research Use Only

Formula: C₁₃H₈BrNO
MW: 274.12 g/mol
MDL Number: MFCD00168898
CAS Number: 3164-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR