Furan-2,5-diyldimethanamine 95%

Supplier: Ambeed
Danger

A564186-10G A564186-250MG A564186-25G A564186-5G
77242-134EA 322.39 USD
77242-134 77242-136 77242-138 77242-140
Furan-2,5-diyldimethanamine 95%
Furan-2,5-diyldimethanamine

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O
MW: 126.15 g/mol
MDL Number: MFCD06212942
CAS Number: 2213-51-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR