VAS-2870 98+%

Supplier: Ambeed
Warning

Synonyms: 3-Benzyl-7-(2-benzoxazolyl)thio-1,2,3-triazolo(4,5-d)pyrimidine, VAS2870, 2-((3-Benzyl-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-yl)thio)benzo[d]oxazole

A564124-10MG A564124-1MG A564124-25MG A564124-5MG
77266-378EA 46 USD
77266-378 77266-380 77266-382 77266-384
VAS-2870 98+%
VAS-2870

Precaución: for Research Use Only

Formula: C₁₈H₁₂N₆OS
MW: 360.4 g/mol
MDL Number: MFCD03407188
CAS Number: 722456-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR