Tetrakis(4-(1H-imidazol-1-yl)phenyl)methane 97%

Supplier: Ambeed
Warning

A563586-100MG A563586-1G A563586-250MG
77266-372EA 67.98 USD
77266-372 77266-374 77266-376
Tetrakis(4-(1H-imidazol-1-yl)phenyl)methane 97%
Tetrakis(4-(1H-imidazol-1-yl)phenyl)methane

Precaución: for Research Use Only

Formula: C₃₇H₂₈N₈
MW: 584.67 g/mol
MDL Number: MFCD28559404
CAS Number: 1258859-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR