3-Hydroxy-4-methyl-2-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A562887-100MG A562887-1G A562887-250MG
77242-114EA 23 USD
77242-114 77242-116 77242-118
3-Hydroxy-4-methyl-2-nitrobenzoic acid 97%
3-Hydroxy-4-methyl-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MDL Number: MFCD00007106
CAS Number: 6946-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR