ω-Bromo-2-hydroxyacetophenone 96%

Supplier: Ambeed
Danger

Synonyms: 2-Hydroxyphenacyl bromide, 2-Bromo-1-(2-hydroxyphenyl)ethan-1-one, 2-Bromo-1-(2-hydroxyphenyl)ethanone, α-Bromo-2-hydroxyacetophenone, 2-Bromo-2'-hydroxyacetophenone

A562878-100G A562878-1G A562878-25G A562878-5G
77242-106EA 483.07 USD
77242-106 77242-108 77242-110 77242-112
ω-Bromo-2-hydroxyacetophenone 96%
ω-Bromo-2-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
MDL Number: MFCD01727570
CAS Number: 2491-36-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR