(S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-(4,4',6,6'-Tetramethoxybiphenyl-2,2'-diyl)bis(di-p-tolylphosphine), (S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxy)-1,1'-biphenyl, (S)-Tolyl-Garphos
A562572-100MG
A562572-250MG
77253-994EA
80.34
USD
77253-994
77253-996
(S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl 98%
(S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl
Precaución: for Research Use Only
Formula:
C₄₄H₄₄O₄P₂ MW: 698.77 g/mol |
MDL Number:
MFCD19443623 CAS Number: 1365531-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...