2-Amino-4,5-dichlorobenzonitrile 97%

Supplier: Ambeed
Warning

A562420-100MG A562420-1G A562420-250MG
77242-100EA 9 USD
77242-100 77242-102 77242-104
2-Amino-4,5-dichlorobenzonitrile 97%
2-Amino-4,5-dichlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
MDL Number: MFCD20695857
CAS Number: 28568-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR