Diacetato[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]ruthenium(II) 98%

Supplier: Ambeed
Warning

Synonyms: Ru(OAc)2[(R)-segphos®]

A562280-1G A562280-250MG A562280-5G
77253-988EA 107 USD
77253-988 77253-990 77253-992
Diacetato[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]ruthenium(II) 98%
Diacetato[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]ruthenium(II)

Precaución: for Research Use Only

Formula: C₄₂H₃₄O₈P₂Ru
MW: 829.7300 g/mol
MDL Number: MFCD09753024
CAS Number: 944450-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR