1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-ol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole

A562067-1G A562067-250MG A562067-5G
77242-084EA 27.81 USD
77242-084 77242-086 77242-088
1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-ol 98%
1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-ol

Precaución: for Research Use Only

Formula: C₅H₅F₃N₂O
MW: 166.1 g/mol
MDL Number: MFCD00139833
CAS Number: 122431-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR