Dihydridotetrakis(triphenylphosphine)ruthenium(II) 98%

Supplier: Ambeed
Warning

A561661-100MG A561661-1G A561661-250MG
77253-982EA 35 USD
77253-982 77253-984 77253-986
Dihydridotetrakis(triphenylphosphine)ruthenium(II) 98%
Dihydridotetrakis(triphenylphosphine)ruthenium(II)

Precaución: for Research Use Only

Formula: C₇₂H₆₀P₄Ru
MW: 1150.237 g/mol
MDL Number: MFCD04126429
CAS Number: 19529-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR