1-Methylimidazole-4-carboxylic acid methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methyl-1H-imidazole-4-carboxylate

A561588-100G A561588-1G A561588-25G A561588-5G
77242-076EA 1184.5 USD
77242-076 77242-078 77242-080 77242-082
1-Methylimidazole-4-carboxylic acid methyl ester 98%
1-Methylimidazole-4-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MW: 140.1400 g/mol
MDL Number: MFCD02179561
CAS Number: 17289-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR