3,4-Dichlorophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3,4-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,4-Dichlorophenylboronic acid, pinacol ester

A561584-1G A561584-250MG A561584-5G
77280-042EA 42.23 USD
77280-042 77280-044 77280-046
3,4-Dichlorophenylboronic acid pinacol ester 98%
3,4-Dichlorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BCl₂O₂
MW: 272.97 g/mol
MDL Number: MFCD05663853
CAS Number: 401797-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR