2,2-Difluoroethylamine hydrochloride 98%

Supplier: Ambeed
Warning

A561550-100G A561550-1G A561550-5G A561550-25G
77253-978EA 151 USD
77253-978 77253-980 77569-084 77572-776
2,2-Difluoroethylamine hydrochloride 98%
2,2-Difluoroethylamine hydrochloride

Precaución: For research use only.

Formula: C₂H₆ClF₂N
MDL Number: MFCD04038283
CAS Number: 79667-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR