Methyl 6-(ethylamino)-5-nitronicotinate 98%

Supplier: Ambeed
Warning

A561529-100MG A561529-1G A561529-250MG A561529-5G
77242-068EA 8 USD
77242-068 77242-070 77242-072 77242-074
Methyl 6-(ethylamino)-5-nitronicotinate 98%
Methyl 6-(ethylamino)-5-nitronicotinate

Precaución: for Research Use Only

Formula: C₉H₁₁N₃O₄
MDL Number: MFCD09862690
CAS Number: 954228-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR