4'-Methoxyacetophenone 95%

Supplier: Ambeed
Warning

A561402-100G A561402-25G A561402-500G
77292-536EA 17 USD
77292-536 77292-538 77292-540
4'-Methoxyacetophenone 95%
4'-Methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MDL Number: MFCD00008745
CAS Number: 100-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR