4-Cyano-3-pyridineboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isonicotinonitrile, 4-Cyanopyridine-3-boronic acid, pinacol ester, 4-Cyanopyridine-3-boronic acid pinacol ester

A561338-100MG A561338-250MG A561338-1G
77292-532EA 18 USD
77292-532 77292-534 77749-159
4-Cyano-3-pyridineboronic acid pinacol ester 96%
4-Cyano-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BN₂O₂
MDL Number: MFCD08458478
CAS Number: 878194-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR