1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate 98%

Supplier: Ambeed
Danger

Synonyms: 1-Butyl-2,3-dimethylimidazolium Tetrafluoroborate, [BDiMIM] [BF4]

A561259-25G A561259-5G A561259-10G
77242-064EA 41 USD
77242-064 77242-066 77747-156
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate 98%
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate

Precaución: for Research Use Only

Formula: C₉H₁₇BF₄N₂
MDL Number: MFCD03427618
CAS Number: 402846-78-0
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR