(4-(tert-Butyl)phenyl)hydrazine xhydrochloride 95%

Supplier: Ambeed
Warning

A560980-1G A560980-250MG A560980-25G A560980-5G
77253-970EA 19 USD
77253-970 77253-972 77253-974 77253-976
(4-(tert-Butyl)phenyl)hydrazine xhydrochloride 95%
(4-(tert-Butyl)phenyl)hydrazine xhydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₇ClN₂
MW: 200.70 g/mol
MDL Number: MFCD00013387
CAS Number: 36600-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR